The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291726.1 | NP_001278655.1 | 6 - 7 | 805 | 81 |
NM_001291728.1 | NP_001278657.1 | 6 - 7 | 895 | 81 | |
NM_033419.4 | NP_219487.3 | 7 - 8 | 958 | 81 | |
XM_011525481.2 | XP_011523783.1 | 8 - 9 | 1087 | 81 | |
GenBank mRNA | AB088396.1 | - | 7 - 8 | 910 | 81 |
AF217980.1 | - | 4 - 5 | 353 | 81 | |
AK057911.1 | - | 7 - 8 | 1266 | 81 | |
AK075474.1 | - | 6 - 7 | 792 | 81 | |
AK294639.1 | - | 6 - 7 | 882 | 81 | |
AK301106.1 | - | 8 - 9 | 860 | 81 | |
AK314469.1 | - | 7 - 8 | 918 | 81 | |
AY358437.1 | - | 7 - 8 | 934 | 81 | |
BC010652.1 | - | 7 - 8 | 919 | 81 |