Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 12 - 13 2097 59
NM_004592.3 NP_004583.2 12 - 13 2097 59
XM_011538654.2 XP_011536956.1 12 - 13 2049 59
XM_011538655.2 XP_011536957.1 12 - 13 2094 59
XM_017019799.1 XP_016875288.1 12 - 13 2536 59
GenBank mRNA AB208921.1 - 10 - 11 5288 59
BC008707.2 - 12 - 13 2094 59
BC015953.2 - 12 - 13 2094 59
BC136678.1 - 12 - 13 2047 59
BC144364.1 - 12 - 13 2047 59
U08377.1 - 12 - 13 2074 59

View Full Product Details