Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 2 - 3 528 109
NM_004592.3 NP_004583.2 2 - 3 528 109
XM_011538653.1 XP_011536955.1 2 - 3 525 109
XM_011538655.2 XP_011536957.1 2 - 3 525 109
XM_017019798.1 XP_016875287.1 2 - 3 525 109
GenBank mRNA BC008707.2 - 2 - 3 525 109
BC015953.2 - 2 - 3 525 109
BC024041.1 - 2 - 3 526 109
BC136678.1 - 2 - 3 478 109
BC144364.1 - 2 - 3 478 109
U08377.1 - 2 - 3 505 109

View Full Product Details