The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261411.1 | NP_001248340.1 | 3 - 4 | 659 | 140 |
NM_004592.3 | NP_004583.2 | 3 - 4 | 659 | 140 | |
XM_011538653.1 | XP_011536955.1 | 3 - 4 | 656 | 140 | |
XM_011538654.2 | XP_011536956.1 | 3 - 4 | 611 | 140 | |
XM_011538655.2 | XP_011536957.1 | 3 - 4 | 656 | 140 | |
XM_017019798.1 | XP_016875287.1 | 3 - 4 | 656 | 140 | |
XM_017019799.1 | XP_016875288.1 | 3 - 4 | 1098 | 140 | |
GenBank mRNA | AB208921.1 | - | 1 - 2 | 3850 | 140 |
BC008707.2 | - | 3 - 4 | 656 | 140 | |
BC015953.2 | - | 3 - 4 | 656 | 140 | |
BC024041.1 | - | 3 - 4 | 657 | 140 | |
BC113420.1 | - | 3 - 4 | 562 | 140 | |
BC136678.1 | - | 3 - 4 | 609 | 140 | |
BC144364.1 | - | 3 - 4 | 609 | 140 | |
U08377.1 | - | 3 - 4 | 636 | 140 |