The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102397.2 | NP_001095867.1 | 4 - 5 | 490 | 79 |
NM_001102398.2 | NP_001095868.1 | 5 - 6 | 656 | 79 | |
NM_001102399.2 | NP_001095869.1 | 4 - 5 | 490 | 79 | |
NM_005826.4 | NP_005817.1 | 5 - 6 | 656 | 79 | |
XM_005245711.4 | XP_005245768.1 | 5 - 6 | 578 | 79 | |
XM_011540471.2 | XP_011538773.1 | 5 - 6 | 578 | 79 | |
GenBank mRNA | AF000364.1 | - | 5 - 6 | 554 | 79 |
AK313142.1 | - | 5 - 6 | 604 | 79 | |
BC001449.2 | - | 5 - 6 | 602 | 79 | |
BC110389.1 | - | 4 - 5 | 417 | 79 | |
BX640912.1 | - | 4 - 5 | 426 | 79 | |
BX647784.1 | - | 4 - 5 | 388 | 79 | |
DA496215.1 | - | 4 - 5 | 490 | 79 | |
DA773186.1 | - | 4 - 5 | 486 | 79 |