The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020205.3 | NP_064590.2 | 3 - 4 | 634 | 62 |
XM_011509782.1 | XP_011508084.1 | 3 - 4 | 631 | 62 | |
XM_011509783.2 | XP_011508085.1 | 3 - 4 | 861 | 62 | |
XM_011509784.2 | XP_011508086.1 | 3 - 4 | 425 | 62 | |
XM_011509785.2 | XP_011508087.1 | 3 - 4 | 4058 | 62 | |
XM_017001851.1 | XP_016857340.1 | 2 - 3 | 483 | 62 | |
GenBank mRNA | AJ293573.1 | - | 3 - 4 | 477 | 62 |
AK299790.1 | - | 3 - 4 | 620 | 62 | |
BC020622.1 | - | 3 - 4 | 420 | 62 | |
BC072681.1 | - | 3 - 4 | 571 | 62 |