The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122772.2 | NP_001116244.1 | 12 - 13 | 2426 | 67 |
NM_014770.3 | NP_055585.1 | 12 - 13 | 1508 | 67 | |
XM_005268625.3 | XP_005268682.1 | 12 - 13 | 2552 | 67 | |
XM_005268626.2 | XP_005268683.1 | 12 - 13 | 1548 | 67 | |
XM_017018772.1 | XP_016874261.1 | 12 - 13 | 2553 | 67 | |
GenBank mRNA | AF384128.1 | - | 12 - 13 | 1504 | 67 |
AF413077.1 | - | 12 - 13 | 1418 | 67 | |
AK122827.1 | - | 12 - 13 | 1509 | 67 | |
AK292672.1 | - | 12 - 13 | 1480 | 67 | |
AY128689.1 | - | 12 - 13 | 2426 | 67 | |
BC028020.1 | - | 12 - 13 | 1475 | 67 | |
D79989.1 | - | 12 - 13 | 1506 | 67 |