Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGAP2
Entrez Gene ID
116986
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTG1, GGAP2, PIKE
Chromosome Location
Chr.12: 57723765 - 57742201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122772.2 NP_001116244.1 18 - 19 3339 98
NM_014770.3 NP_055585.1 17 - 18 2361 98
XM_005268625.3 XP_005268682.1 17 - 18 3405 98
XM_005268626.2 XP_005268683.1 18 - 19 2461 98
XM_017018772.1 XP_016874261.1 18 - 19 3466 98
GenBank mRNA AF384128.1 - 17 - 18 2357 98
AF413077.1 - 17 - 18 2271 98
AK122827.1 - 18 - 19 2422 98
AK292672.1 - 17 - 18 2333 98
AY128689.1 - 18 - 19 3339 98
BC028020.1 - 17 - 18 2328 98
D79989.1 - 17 - 18 2359 98

View Full Product Details