The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000103.3 | NP_000094.2 | 2 - 3 | 293 | 126 |
NM_031226.2 | NP_112503.1 | 3 - 4 | 402 | 126 | |
XM_005254191.1 | XP_005254248.1 | 2 - 3 | 250 | 126 | |
GenBank mRNA | AK291778.1 | - | 3 - 4 | 400 | 126 |
BC020767.1 | - | 2 - 3 | 291 | 126 | |
BC022896.1 | - | 4 - 5 | 630 | 126 | |
BC035714.1 | - | 1 - 2 | 250 | 126 | |
BC035959.1 | - | 2 - 3 | 290 | 126 | |
BC107785.1 | - | 2 - 3 | 295 | 126 | |
DQ118405.1 | - | 2 - 3 | 234 | 126 | |
J04127.1 | - | 1 - 2 | 178 | 126 | |
M22246.1 | - | 2 - 3 | 275 | 126 | |
X13589.1 | - | 2 - 3 | 275 | 126 | |
Y07508.1 | - | 1 - 2 | 200 | 126 |