The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052950.3 | NP_443182.1 | 1 - 2 | 476 | 72 |
XM_011534914.1 | XP_011533216.1 | 1 - 2 | 367 | 72 | |
XM_011534915.2 | XP_011533217.1 | 1 - 2 | 630 | 72 | |
GenBank mRNA | AF411978.1 | - | 1 - 2 | 136 | 72 |
AK309815.1 | - | 1 - 2 | 329 | 72 | |
AK311177.1 | - | 1 - 2 | 362 | 72 | |
BC014004.2 | - | 1 - 2 | 316 | 72 | |
DA124107.1 | - | 1 - 2 | 476 | 72 |