The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052950.3 | NP_443182.1 | 4 - 5 | 674 | 136 |
XM_011534914.1 | XP_011533216.1 | 4 - 5 | 565 | 136 | |
GenBank mRNA | AK309815.1 | - | 4 - 5 | 527 | 136 |
BC014004.2 | - | 4 - 5 | 514 | 136 |