Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCORL1
Entrez Gene ID
63035
Gene Name
BCL6 corepressor-like 1
Gene Aliases
BCoR-L1, CXorf10
Chromosome Location
Chr.X: 129980285 - 130058083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005262456.4 XP_005262513.2 7 - 8 4149 83
GenBank mRNA AL096777.1 - 3 - 4 1995 83

View Full Product Details