The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020718.3 | NP_065769.3 | 8 - 9 | 1529 | 58 |
XM_017023496.1 | XP_016878985.1 | 8 - 9 | 1560 | 58 | |
XM_017023497.1 | XP_016878986.1 | 8 - 9 | 3055 | 58 | |
XM_017023498.1 | XP_016878987.1 | 7 - 8 | 842 | 58 | |
XM_017023499.1 | XP_016878988.1 | 5 - 6 | 646 | 58 | |
XM_017023500.1 | XP_016878989.1 | 8 - 9 | 1560 | 58 | |
GenBank mRNA | AJ586135.2 | - | 8 - 9 | 1529 | 58 |
AK126752.1 | - | 8 - 9 | 1213 | 58 | |
BM842261.1 | - | 2 - 3 | 226 | 58 |