The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321378.1 | NP_001308307.1 | 27 - 28 | 4395 | 69 |
NM_001321380.1 | NP_001308309.1 | 25 - 26 | 3196 | 69 | |
NM_002645.3 | NP_002636.2 | 26 - 27 | 4326 | 69 | |
XM_011520186.2 | XP_011518488.1 | 25 - 26 | 4153 | 69 | |
XM_017017924.1 | XP_016873413.1 | 27 - 28 | 4342 | 69 | |
GenBank mRNA | AK294416.1 | - | 25 - 26 | 3183 | 69 |
AK309610.1 | - | 7 - 8 | 759 | 69 | |
BC113658.1 | - | 25 - 26 | 4178 | 69 | |
BX648778.1 | - | 16 - 17 | 2262 | 69 | |
Y13367.1 | - | 25 - 26 | 4112 | 69 |