Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135214.1 15 - 16 2644 98
NR_135215.1 15 - 16 2626 98
NR_135216.1 15 - 16 2531 98
NR_135217.1 14 - 15 2341 98
GenBank mRNA AF416901.1 - 15 - 16 2490 98
AF416902.1 - 14 - 15 2205 98
AF416903.1 - 15 - 16 2508 98
AF416904.1 - 14 - 15 2724 98
AF416905.1 - 15 - 16 2395 98
AK301257.1 - 14 - 15 2747 98

View Full Product Details