Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
Chromosome Location
Chr.1: 157676481 - 157700985 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135214.1 7 - 8 1437 61
GenBank mRNA AF416903.1 - 7 - 8 1301 61

View Full Product Details