The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018016.2 | NP_001018016.1 | 2 - 3 | 251 | 66 |
NM_001018017.2 | NP_001018017.1 | 2 - 3 | 224 | 66 | |
NM_001044391.2 | NP_001037856.1 | 2 - 3 | 224 | 66 | |
NM_001044392.2 | NP_001037857.1 | 2 - 3 | 251 | 66 | |
NM_001204288.1 | NP_001191217.1 | 2 - 3 | 251 | 66 | |
NM_001204295.1 | NP_001191224.1 | 2 - 3 | 251 | 66 | |
GenBank mRNA | AF125525.1 | - | 2 - 3 | 143 | 66 |
AY327583.1 | - | 2 - 3 | 176 | 66 | |
AY327584.1 | - | 2 - 3 | 203 | 66 | |
AY327587.1 | - | 2 - 3 | 229 | 66 | |
AY327595.1 | - | 2 - 3 | 176 | 66 | |
AY327600.1 | - | 2 - 3 | 176 | 66 | |
BC120974.2 | - | 2 - 3 | 201 | 66 | |
BC120975.2 | - | 2 - 3 | 228 | 66 | |
EF583645.1 | - | 2 - 3 | 328 | 66 | |
EF583649.1 | - | 2 - 3 | 229 | 66 | |
EF583650.1 | - | 2 - 3 | 166 | 66 | |
EF583651.1 | - | 2 - 3 | 166 | 66 | |
EF583655.1 | - | 2 - 3 | 229 | 66 | |
EF670710.1 | - | 2 - 3 | 202 | 66 | |
EF670713.1 | - | 2 - 3 | 229 | 66 | |
EF670716.1 | - | 2 - 3 | 229 | 66 | |
EF670717.1 | - | 2 - 3 | 229 | 66 | |
FJ226032.1 | - | 1 - 2 | 704 | 66 | |
FJ226038.1 | - | 1 - 2 | 703 | 66 | |
FJ226040.1 | - | 2 - 3 | 229 | 66 | |
FJ226052.1 | - | 2 - 3 | 202 | 66 | |
U60260.1 | - | 2 - 3 | 177 | 66 | |
X80761.1 | - | 2 - 3 | 152 | 66 |