The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324475.1 | NP_001311404.1 | 4 - 5 | 549 | 75 |
NM_023015.4 | NP_075391.3 | 3 - 4 | 890 | 75 | |
XM_005245459.2 | XP_005245516.1 | 3 - 4 | 907 | 75 | |
XM_006711490.3 | XP_006711553.1 | 4 - 5 | 360 | 75 | |
XM_011509906.2 | XP_011508208.1 | 3 - 4 | 272 | 75 | |
GenBank mRNA | AK290025.1 | - | 4 - 5 | 543 | 75 |
AL832133.1 | - | 3 - 4 | 274 | 75 | |
BC105092.1 | - | 3 - 4 | 353 | 75 | |
BC105094.1 | - | 3 - 4 | 353 | 75 | |
BC116458.1 | - | 3 - 4 | 885 | 75 | |
BX640786.1 | - | 3 - 4 | 876 | 75 | |
BX640950.1 | - | 3 - 4 | 891 | 75 | |
CR627233.1 | - | 3 - 4 | 890 | 75 | |
CR749212.1 | - | 3 - 4 | 884 | 75 | |
CR749376.1 | - | 3 - 4 | 891 | 75 |