Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
Chromosome Location
Chr.X: 108155607 - 108439497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287758.1 NP_001274687.1 14 - 15 1144 77
NM_001287759.1 NP_001274688.1 14 - 15 1144 77
NM_001287760.1 NP_001274689.1 14 - 15 1144 77
NM_001847.3 NP_001838.2 14 - 15 1039 77
NM_033641.3 NP_378667.1 14 - 15 1144 77
XM_006724617.3 XP_006724680.1 14 - 15 1033 77
XM_011530852.2 XP_011529154.1 14 - 15 1033 77
XM_011530853.2 XP_011529155.1 12 - 13 1749 77
XM_011530854.2 XP_011529156.1 14 - 15 1034 77
GenBank mRNA BC140855.1 - 14 - 15 1040 77
BC144669.1 - 14 - 15 1040 77
D21337.1 - 14 - 15 1144 77
L22763.1 - 14 - 15 1022 77
U04845.1 - 14 - 15 1022 77

View Full Product Details