Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MYC
- Entrez Gene ID
-
4609
- Gene Name
- v-myc avian myelocytomatosis viral oncogene homolog
- Gene Aliases
-
MRTL, MYCC, bHLHe39, c-Myc
- Chromosome Location
-
Chr.8: 127736069 - 127741434 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M13929.1 |
- |
2 - 3 |
503 |
84 |
View Full Product Details