The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014109.3 | NP_054828.2 | 4 - 5 | 646 | 83 |
XM_011516994.2 | XP_011515296.1 | 4 - 5 | 1173 | 83 | |
XM_011516995.2 | XP_011515297.1 | 4 - 5 | 1173 | 83 | |
GenBank mRNA | AK097133.1 | - | 4 - 5 | 628 | 83 |
AL833653.1 | - | 4 - 5 | 400 | 83 | |
AY598335.1 | - | 4 - 5 | 538 | 83 | |
BC113656.1 | - | 4 - 5 | 560 | 83 | |
CR749832.1 | - | 5 - 6 | 902 | 83 |