The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020429.2 | NP_065162.1 | 17 - 18 | 2425 | 109 |
NM_181349.2 | NP_851994.1 | 16 - 17 | 2347 | 109 | |
GenBank mRNA | AB046845.1 | - | 17 - 18 | 2393 | 109 |
AF199364.2 | - | 16 - 17 | 1980 | 109 | |
AF464850.1 | - | 17 - 18 | 2085 | 109 | |
BC152468.1 | - | 17 - 18 | 2393 | 109 |