The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278185.1 | NP_001265114.1 | 18 - 19 | 2360 | 93 |
NM_001278186.1 | NP_001265115.1 | 15 - 16 | 2084 | 93 | |
NM_015601.3 | NP_056416.2 | 17 - 18 | 2336 | 93 | |
NM_022079.2 | NP_071362.1 | 18 - 19 | 2360 | 93 | |
XM_011539592.2 | XP_011537894.1 | 19 - 20 | 2434 | 93 | |
XM_011539593.2 | XP_011537895.1 | 18 - 19 | 2410 | 93 | |
XM_011539594.2 | XP_011537896.1 | 20 - 21 | 2520 | 93 | |
XM_011539595.2 | XP_011537897.1 | 20 - 21 | 2570 | 93 | |
XM_011539596.2 | XP_011537898.1 | 19 - 20 | 2448 | 93 | |
XM_011539597.2 | XP_011537899.1 | 17 - 18 | 2071 | 93 | |
XM_017016041.1 | XP_016871530.1 | 16 - 17 | 1999 | 93 | |
GenBank mRNA | AB046813.1 | - | 15 - 16 | 2214 | 93 |
AK027835.1 | - | 8 - 9 | 903 | 93 | |
AK292809.1 | - | 17 - 18 | 2216 | 93 | |
AY221963.1 | - | 18 - 19 | 2297 | 93 | |
AY650032.1 | - | 15 - 16 | 2021 | 93 | |
AY650033.1 | - | 18 - 19 | 2297 | 93 | |
BC015043.1 | - | 15 - 16 | 1987 | 93 | |
BC039600.1 | - | 17 - 18 | 2246 | 93 | |
BX537563.1 | - | 17 - 18 | 4823 | 93 |