The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032961.2 | NP_116586.1 | 1 - 2 | 3484 | 79 |
XM_011532150.1 | XP_011530452.1 | 1 - 2 | 3484 | 79 | |
GenBank mRNA | AB037821.1 | - | 1 - 2 | 3459 | 79 |
AK024636.1 | - | 1 - 2 | 748 | 79 | |
AL157470.1 | - | 1 - 2 | 541 | 79 | |
BC111560.1 | - | 1 - 2 | 2822 | 79 | |
KJ534913.1 | - | 1 - 2 | 2633 | 79 | |
KJ534914.1 | - | 2 - 3 | 2462 | 79 |