Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTM1
Entrez Gene ID
28962
Gene Name
osteopetrosis associated transmembrane protein 1
Gene Aliases
GIPN, GL, HSPC019, OPTB5
Chromosome Location
Chr.6: 108041409 - 108074737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014028.3 NP_054747.2 1 - 2 483 68
GenBank mRNA AF533891.1 - 1 - 2 467 68
AK075012.1 - 1 - 2 409 68
AK129854.1 - 1 - 2 477 68
AK314393.1 - 1 - 2 159 68
AY358795.1 - 1 - 2 474 68
BC068581.1 - 1 - 2 465 68
BI668275.1 - 1 - 2 500 68

View Full Product Details