The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290185.1 | NP_001277114.1 | 12 - 13 | 2349 | 83 |
NM_021729.5 | NP_068375.3 | 12 - 13 | 2134 | 83 | |
XM_011542898.1 | XP_011541200.1 | 9 - 10 | 1367 | 83 | |
XM_011542899.2 | XP_011541201.1 | 12 - 13 | 2233 | 83 | |
GenBank mRNA | AB027508.2 | - | 12 - 13 | 2104 | 83 |
AF258558.1 | - | 10 - 11 | 1634 | 83 | |
AF308800.1 | - | 12 - 13 | 2068 | 83 | |
AK023279.1 | - | 12 - 13 | 2105 | 83 | |
AK026012.1 | - | 3 - 4 | 340 | 83 | |
AK223158.1 | - | 12 - 13 | 2094 | 83 | |
AK302973.1 | - | 12 - 13 | 2281 | 83 | |
AL834218.1 | - | 12 - 13 | 2081 | 83 | |
BC012051.1 | - | 4 - 5 | 647 | 83 | |
BC065563.1 | - | 12 - 13 | 2068 | 83 |