The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018054.2 | NP_001018064.1 | 5 - 6 | 1122 | 80 |
NM_004631.4 | NP_004622.2 | 5 - 6 | 1122 | 80 | |
XM_005271173.3 | XP_005271230.1 | 5 - 6 | 1032 | 80 | |
XM_006710881.3 | XP_006710944.1 | 5 - 6 | 1032 | 80 | |
XM_006710882.3 | XP_006710945.1 | 5 - 6 | 1032 | 80 | |
XM_011542094.2 | XP_011540396.1 | 5 - 6 | 1032 | 80 | |
XM_011542096.2 | XP_011540398.1 | 5 - 6 | 1032 | 80 | |
XM_017002265.1 | XP_016857754.1 | 5 - 6 | 1032 | 80 | |
XM_017002266.1 | XP_016857755.1 | 5 - 6 | 1032 | 80 | |
GenBank mRNA | BC051836.1 | - | 5 - 6 | 1027 | 80 |
D50678.1 | - | 5 - 6 | 987 | 80 |