The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142550.1 | NP_001136022.1 | 10 - 11 | 2326 | 64 |
NM_001142551.1 | NP_001136023.1 | 10 - 11 | 2302 | 64 | |
NM_014969.5 | NP_055784.3 | 10 - 11 | 2305 | 64 | |
XM_006710460.3 | XP_006710523.1 | 10 - 11 | 2297 | 64 | |
XM_011541027.2 | XP_011539329.1 | 10 - 11 | 2315 | 64 | |
XM_011541028.2 | XP_011539330.1 | 10 - 11 | 2294 | 64 | |
XM_011541029.1 | XP_011539331.1 | 10 - 11 | 2318 | 64 | |
XM_011541030.1 | XP_011539332.1 | 10 - 11 | 1966 | 64 | |
XM_011541031.1 | XP_011539333.1 | 8 - 9 | 1793 | 64 | |
XM_017000696.1 | XP_016856185.1 | 10 - 11 | 2315 | 64 | |
XM_017000697.1 | XP_016856186.1 | 10 - 11 | 1963 | 64 | |
GenBank mRNA | AB020700.1 | - | 10 - 11 | 2149 | 64 |
AK074616.1 | - | 3 - 4 | 329 | 64 | |
AK225781.1 | - | 10 - 11 | 2086 | 64 | |
AK289789.1 | - | 10 - 11 | 2179 | 64 | |
AK294998.1 | - | 7 - 8 | 1698 | 64 | |
BC034964.2 | - | 10 - 11 | 2190 | 64 | |
BC039254.1 | - | 9 - 10 | 2092 | 64 |