The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190879.2 | NP_001177808.1 | 13 - 14 | 1338 | 173 |
NM_001190881.2 | NP_001177810.1 | 9 - 10 | 956 | 173 | |
NM_001190882.2 | NP_001177811.1 | 11 - 12 | 1100 | 173 | |
NM_002552.4 | NP_002543.2 | 12 - 13 | 1222 | 173 | |
NM_181741.3 | NP_859525.1 | 12 - 13 | 1331 | 173 | |
NM_181742.3 | NP_859526.1 | 12 - 13 | 1226 | 173 | |
XM_011511255.2 | XP_011509557.1 | 13 - 14 | 1376 | 173 | |
XM_017004226.1 | XP_016859715.1 | 14 - 15 | 1436 | 173 | |
XM_017004227.1 | XP_016859716.1 | 11 - 12 | 1060 | 173 | |
GenBank mRNA | AF022108.1 | - | 12 - 13 | 1178 | 173 |
AF047598.1 | - | 12 - 13 | 1177 | 173 | |
AF132596.1 | - | 12 - 13 | 1164 | 173 | |
AK225092.1 | - | 12 - 13 | 1139 | 173 | |
AK291989.1 | - | 12 - 13 | 1196 | 173 | |
AK295721.1 | - | 9 - 10 | 925 | 173 | |
AK298862.1 | - | 11 - 12 | 1100 | 173 | |
AK299749.1 | - | 10 - 11 | 961 | 173 | |
BC005388.1 | - | 12 - 13 | 1143 | 173 | |
BC014847.1 | - | 12 - 13 | 1165 | 173 | |
BC016177.1 | - | 8 - 9 | 814 | 173 |