The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271751.1 | NP_001258680.1 | 12 - 13 | 1513 | 157 |
NM_005795.5 | NP_005786.1 | 13 - 14 | 1678 | 157 | |
XM_005246231.3 | XP_005246288.2 | 14 - 15 | 1722 | 157 | |
XM_005246232.3 | XP_005246289.2 | 15 - 16 | 1763 | 157 | |
XM_005246234.3 | XP_005246291.2 | 13 - 14 | 1594 | 157 | |
XM_017003151.1 | XP_016858640.1 | 14 - 15 | 1653 | 157 | |
GenBank mRNA | AK291630.1 | - | 13 - 14 | 1636 | 157 |
AK292998.1 | - | 12 - 13 | 1471 | 157 | |
AY389506.1 | - | 10 - 11 | 1130 | 157 | |
CB270158.1 | - | 1 - 2 | 71 | 157 | |
L76380.1 | - | 13 - 14 | 1684 | 157 | |
U17473.1 | - | 13 - 14 | 1600 | 157 |