The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271751.1 | NP_001258680.1 | 6 - 7 | 789 | 133 |
NM_005795.5 | NP_005786.1 | 7 - 8 | 954 | 133 | |
XM_005246231.3 | XP_005246288.2 | 8 - 9 | 998 | 133 | |
XM_005246232.3 | XP_005246289.2 | 9 - 10 | 1039 | 133 | |
XM_005246234.3 | XP_005246291.2 | 7 - 8 | 870 | 133 | |
XM_017003151.1 | XP_016858640.1 | 8 - 9 | 929 | 133 | |
GenBank mRNA | AK291630.1 | - | 7 - 8 | 912 | 133 |
AK292998.1 | - | 6 - 7 | 747 | 133 | |
AY389506.1 | - | 4 - 5 | 406 | 133 | |
L76380.1 | - | 7 - 8 | 960 | 133 | |
U17473.1 | - | 7 - 8 | 876 | 133 |