Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
Chromosome Location
Chr.2: 187341963 - 187448307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271751.1 NP_001258680.1 7 - 8 881 125
NM_005795.5 NP_005786.1 8 - 9 1046 125
XM_005246231.3 XP_005246288.2 9 - 10 1090 125
XM_005246232.3 XP_005246289.2 10 - 11 1131 125
XM_005246234.3 XP_005246291.2 8 - 9 962 125
XM_017003151.1 XP_016858640.1 9 - 10 1021 125
GenBank mRNA AK291630.1 - 8 - 9 1004 125
AK292998.1 - 7 - 8 839 125
AY389506.1 - 5 - 6 498 125
L76380.1 - 8 - 9 1052 125
U17473.1 - 8 - 9 968 125

View Full Product Details