The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324403.1 | NP_001311332.1 | 2 - 3 | 480 | 62 |
NM_015886.4 | NP_056970.1 | 2 - 3 | 452 | 62 | |
GenBank mRNA | BC074931.2 | - | 1 - 2 | 296 | 62 |
BC074932.2 | - | 1 - 2 | 296 | 62 | |
BC126290.1 | - | 1 - 2 | 301 | 62 | |
BC126292.1 | - | 1 - 2 | 303 | 62 | |
CR749657.1 | - | 2 - 3 | 453 | 62 | |
D45027.1 | - | 1 - 2 | 757 | 62 |