The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006951.4 | NP_008882.2 | 4 - 5 | 1310 | 118 |
NM_139052.2 | NP_620640.1 | 4 - 5 | 1310 | 118 | |
GenBank mRNA | AK223615.1 | - | 4 - 5 | 1282 | 118 |
AK291229.1 | - | 4 - 5 | 1310 | 118 | |
BC052268.1 | - | 4 - 5 | 1288 | 118 | |
BC136340.1 | - | 4 - 5 | 1291 | 118 | |
BC136348.1 | - | 4 - 5 | 1291 | 118 | |
U80191.1 | - | 4 - 5 | 1287 | 118 | |
X95525.1 | - | 4 - 5 | 1296 | 118 |