Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEP15
Entrez Gene ID
9403
Gene Name
15 kDa selenoprotein
Gene Aliases
-
Chromosome Location
Chr.1: 86862445 - 86914424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004261.3 NP_004252.2 3 - 4 626 81
GenBank mRNA AF051894.1 - 3 - 4 308 81
AF288991.1 - 3 - 4 308 81
AK225252.1 - 3 - 4 317 81
AK225640.1 - 3 - 4 316 81
AL833575.1 - 3 - 4 329 81
BC005294.1 - 3 - 4 334 81
BC016359.1 - 3 - 4 321 81
BQ429619.1 - 3 - 4 341 81

View Full Product Details