Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIK3CA
Entrez Gene ID
5290
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
Gene Aliases
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
Chromosome Location
Chr.3: 179148114 - 179240093 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006218.3 NP_006209.2 18 - 19 2826 71
XM_006713658.3 XP_006713721.1 18 - 19 2851 71
XM_011512894.2 XP_011511196.1 18 - 19 2932 71
GenBank mRNA AB210020.1 - 17 - 18 2674 71
AK297203.1 - 15 - 16 2065 71
AK302608.1 - 17 - 18 2504 71
BC113601.1 - 17 - 18 2746 71
BC113603.1 - 17 - 18 2746 71
U79143.1 - 17 - 18 2669 71
Z29090.1 - 17 - 18 2681 71

View Full Product Details