Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLVAP
Entrez Gene ID
83483
Gene Name
plasmalemma vesicle associated protein
Gene Aliases
FELS, PV-1, PV1, gp68
Chromosome Location
Chr.19: 17351448 - 17377349 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031310.2 NP_112600.1 1 - 2 426 71
GenBank mRNA AF348827.1 - 1 - 2 415 71
AK096030.1 - 1 - 2 419 71
AK313837.1 - 1 - 2 419 71
AL834363.1 - 1 - 2 408 71
BC050365.1 - 1 - 2 405 71
BC056414.1 - 1 - 2 427 71

View Full Product Details