The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031310.2 | NP_112600.1 | 2 - 3 | 531 | 88 |
GenBank mRNA | AF326591.1 | - | 2 - 3 | 522 | 88 |
AF348827.1 | - | 2 - 3 | 520 | 88 | |
AK096030.1 | - | 2 - 3 | 524 | 88 | |
AK313837.1 | - | 2 - 3 | 524 | 88 | |
AL834363.1 | - | 2 - 3 | 513 | 88 | |
BC050365.1 | - | 2 - 3 | 510 | 88 | |
BC056414.1 | - | 2 - 3 | 532 | 88 | |
BX380375.2 | - | 2 - 3 | 532 | 88 |