The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031310.2 | NP_112600.1 | 4 - 5 | 1300 | 97 |
GenBank mRNA | AF326591.1 | - | 4 - 5 | 1291 | 97 |
AF348827.1 | - | 4 - 5 | 1289 | 97 | |
AK074054.1 | - | 2 - 3 | 382 | 97 | |
AK096030.1 | - | 4 - 5 | 1278 | 97 | |
AK313837.1 | - | 5 - 6 | 1221 | 97 | |
AL834363.1 | - | 4 - 5 | 1282 | 97 | |
BC050365.1 | - | 4 - 5 | 1279 | 97 | |
BC056414.1 | - | 4 - 5 | 1301 | 97 |