Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H1A
Entrez Gene ID
3024
Gene Name
histone cluster 1, H1a
Gene Aliases
H1.1, H1A, H1F1, HIST1
Chromosome Location
Chr.6: 26017032 - 26017812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005325.3 NP_005316.1 1 - 1 711 84
GenBank mRNA BC069492.1 - 1 - 1 682 84
BC101593.1 - 1 - 1 710 84
BC112140.1 - 1 - 1 711 84

View Full Product Details