The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320974.1 | NP_001307903.1 | 1 - 2 | 706 | 97 |
NM_001320975.1 | NP_001307904.1 | 1 - 2 | 706 | 97 | |
NM_001320976.1 | NP_001307905.1 | 1 - 2 | 706 | 97 | |
NM_004376.6 | NP_004367.2 | 1 - 2 | 706 | 97 | |
NM_078470.5 | NP_510870.1 | 1 - 2 | 706 | 97 | |
XM_006717634.3 | XP_006717697.1 | 1 - 2 | 171 | 97 | |
XM_011539298.2 | XP_011537600.1 | 1 - 2 | 171 | 97 | |
XM_017015715.1 | XP_016871204.1 | 1 - 2 | 707 | 97 | |
GenBank mRNA | AK291654.1 | - | 1 - 2 | 135 | 97 |
AK298864.1 | - | 1 - 2 | 135 | 97 | |
BC002382.2 | - | 1 - 2 | 105 | 97 | |
BC013403.2 | - | 1 - 2 | 105 | 97 | |
BC078161.1 | - | 1 - 2 | 117 | 97 | |
BT007129.1 | - | 1 - 2 | 89 | 97 | |
BX537557.1 | - | 1 - 2 | 706 | 97 |