Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
Chromosome Location
Chr.2: 135741619 - 135785063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014607.3 NP_055422.1 11 - 12 1497 117
GenBank mRNA AK026971.1 - 11 - 12 1120 117
AK095402.1 - 7 - 8 885 117
AK292829.1 - 11 - 12 1190 117
AK304096.1 - 11 - 12 1207 117
BC035594.2 - 12 - 13 1329 117
BX647216.1 - 9 - 10 2187 117
BX647662.1 - 11 - 12 1289 117
D87684.1 - 11 - 12 1250 117
EF199840.1 - 11 - 12 1314 117

View Full Product Details