Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBXN4
Entrez Gene ID
23190
Gene Name
UBX domain protein 4
Gene Aliases
UBXD2, UBXDC1, erasin
Chromosome Location
Chr.2: 135741619 - 135785063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014607.3 NP_055422.1 3 - 4 527 90
GenBank mRNA AK026971.1 - 3 - 4 150 90
AK292829.1 - 3 - 4 220 90
AK304096.1 - 3 - 4 291 90
BC035594.2 - 4 - 5 359 90
BX647662.1 - 3 - 4 319 90
D87684.1 - 3 - 4 280 90
EF199840.1 - 3 - 4 344 90

View Full Product Details