The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022917.4 | NP_075068.2 | 15 - 16 | 2055 | 104 |
XM_017015044.1 | XP_016870533.1 | 15 - 16 | 2011 | 104 | |
GenBank mRNA | AF361079.1 | - | 15 - 16 | 2013 | 104 |
AF361080.1 | - | 15 - 16 | 2013 | 104 | |
AK056559.1 | - | 7 - 8 | 769 | 104 | |
AK293969.1 | - | 14 - 15 | 104 | ||
BC008852.2 | - | 4 - 5 | 448 | 104 | |
BC030139.2 | - | 15 - 16 | 1959 | 104 |