Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIT2
Entrez Gene ID
56954
Gene Name
nitrilase family member 2
Gene Aliases
HEL-S-8a
Chromosome Location
Chr.3: 100334718 - 100355634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020202.4 NP_064587.1 8 - 9 752 90
GenBank mRNA AF260334.1 - 8 - 9 751 90
AF284574.1 - 8 - 9 699 90
AK225979.1 - 7 - 8 1230 90
AK313704.1 - 8 - 9 694 90
BC020620.1 - 8 - 9 709 90
BC107890.1 - 8 - 9 740 90
FJ224326.1 - 8 - 9 699 90

View Full Product Details