Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIT2
Entrez Gene ID
56954
Gene Name
nitrilase family member 2
Gene Aliases
HEL-S-8a
Chromosome Location
Chr.3: 100334718 - 100355634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020202.4 NP_064587.1 9 - 10 819 70
GenBank mRNA AF260334.1 - 9 - 10 818 70
AF284574.1 - 9 - 10 766 70
AK225979.1 - 8 - 9 1297 70
AK313704.1 - 9 - 10 761 70
BC020620.1 - 9 - 10 776 70
BC107890.1 - 9 - 10 807 70
FJ224326.1 - 9 - 10 766 70

View Full Product Details