Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP4
Entrez Gene ID
7079
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
Chromosome Location
Chr.3: 12153068 - 12159351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003256.3 NP_003247.1 1 - 2 649 73
GenBank mRNA AK313018.1 - 1 - 2 210 73
BC010553.1 - 1 - 2 649 73
BT019627.1 - 1 - 2 138 73
U76456.1 - 1 - 2 197 73

View Full Product Details