The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272073.1 | NP_001259002.1 | 11 - 12 | 1342 | 101 |
NM_005051.2 | NP_005042.1 | 11 - 12 | 1375 | 101 | |
NR_073590.1 | 11 - 12 | 1326 | 101 | ||
XM_017006965.1 | XP_016862454.1 | 11 - 12 | 1055 | 101 | |
GenBank mRNA | AF130067.1 | - | 7 - 8 | 503 | 101 |
AK056815.1 | - | 11 - 12 | 954 | 101 | |
AK222510.1 | - | 11 - 12 | 1003 | 101 | |
AK290523.1 | - | 11 - 12 | 1003 | 101 | |
AK293260.1 | - | 10 - 11 | 902 | 101 | |
AK297987.1 | - | 10 - 11 | 849 | 101 | |
AK301559.1 | - | 11 - 12 | 971 | 101 | |
BC000394.2 | - | 11 - 12 | 984 | 101 | |
BC001567.1 | - | 11 - 12 | 982 | 101 | |
BC001772.1 | - | 7 - 8 | 510 | 101 | |
BC016634.1 | - | 10 - 11 | 807 | 101 | |
BC029739.1 | - | 11 - 12 | 985 | 101 | |
X76013.1 | - | 11 - 12 | 984 | 101 |