The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272073.1 | NP_001259002.1 | 20 - 21 | 2326 | 62 |
NM_005051.2 | NP_005042.1 | 20 - 21 | 2359 | 62 | |
NR_073590.1 | 20 - 21 | 2310 | 62 | ||
XM_017006965.1 | XP_016862454.1 | 20 - 21 | 2039 | 62 | |
GenBank mRNA | AF130067.1 | - | 16 - 17 | 1487 | 62 |
AK056815.1 | - | 20 - 21 | 1938 | 62 | |
AK222510.1 | - | 20 - 21 | 1987 | 62 | |
AK290523.1 | - | 20 - 21 | 1987 | 62 | |
AK293260.1 | - | 19 - 20 | 1886 | 62 | |
AK297987.1 | - | 19 - 20 | 1833 | 62 | |
AK301559.1 | - | 20 - 21 | 1955 | 62 | |
BC000394.2 | - | 20 - 21 | 1968 | 62 | |
BC001567.1 | - | 20 - 21 | 1966 | 62 | |
BC001772.1 | - | 16 - 17 | 1494 | 62 | |
BC016634.1 | - | 19 - 20 | 1791 | 62 | |
BC029739.1 | - | 20 - 21 | 1969 | 62 | |
X76013.1 | - | 20 - 21 | 1968 | 62 |