The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272073.1 | NP_001259002.1 | 4 - 5 | 816 | 80 |
NM_005051.2 | NP_005042.1 | 4 - 5 | 849 | 80 | |
NR_073590.1 | 4 - 5 | 800 | 80 | ||
XM_017006965.1 | XP_016862454.1 | 4 - 5 | 529 | 80 | |
GenBank mRNA | AK056815.1 | - | 4 - 5 | 428 | 80 |
AK222510.1 | - | 4 - 5 | 477 | 80 | |
AK290523.1 | - | 4 - 5 | 477 | 80 | |
AK293260.1 | - | 3 - 4 | 376 | 80 | |
AK297987.1 | - | 3 - 4 | 323 | 80 | |
AK301559.1 | - | 4 - 5 | 445 | 80 | |
AK302867.1 | - | 4 - 5 | 846 | 80 | |
BC000394.2 | - | 4 - 5 | 458 | 80 | |
BC001567.1 | - | 4 - 5 | 456 | 80 | |
BC016634.1 | - | 3 - 4 | 281 | 80 | |
BC029739.1 | - | 4 - 5 | 459 | 80 | |
X76013.1 | - | 4 - 5 | 458 | 80 |